Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Environment
Copper(II) chloride dihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1792-01
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Environment
Copper(II) chloride dihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1792-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 101.0 %
  • ACS - Assay (CuCl₂2H₂O):
    99.0 - 101.0 %
  • ACS - Insoluble Matter:
    ≤ 0.01 %
  • ACS - pH of 5% Solution at 25°C:
    2.0 - 4.0
  • ACS - Nitrate (NO₃):
    ≤ 0.015 %
  • ACS - Sulfate (SO₄):
    ≤ 0.005 %
  • ACS - Calcium (Ca):
    ≤ 0.005 %
  • ACS - Iron (Fe):
    ≤ 0.005 %
  • ACS - Nickel (Ni):
    ≤ 0.01 %
  • ACS - Potassium (K):
    ≤ 0.01 %
  • ACS - Sodium (Na):
    ≤ 0.02 %

Specification Test Results

About this item

 :
 :170.48 g/mol
Merck Index:13,02660
Boiling Pt:1365.85 °C (1013 hPa)
Melting Pt:628 °C
Density:3.054 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
 :GHS07
 :GHS09
CAS Number:10125-13-0
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674