Order Entry
Czech Republic
ContactUsLinkComponent
D-(+)-Sucrose, crystals, BAKER ANALYZED® ACS, J.T.Baker®
D-(+)-Sucrose, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
D-(+)-Sucrose, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    66.3 - 66.8 Degree
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Specific Rotation (+):
    66.3 - 66.8 Degree
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Loss on Drying at 105°C:
    ≤0.03 %
  • ACS - Residue after Ignition:
    ≤0.01 %
  • ACS - Titrable Acid (meq/g):
    ≤0.0008
  • ACS - Chloride (Cl):
    ≤0.005 %
  • ACS - Sulfate and Sulfite (as SO₄):
    ≤0.005 %
  • ACS - Invert Sugar:
    ≤0.05 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together