Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Ammonium chloride, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
 
Ammonium chloride, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Assay (argentometric titrn.):
    min. 99.5 %
  • Calcium (Ca):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.005 %
  • pH of 5% Solution at 25°C:
    4.5 - 5.5
  • Residue after Ignition:
    max. 0.01 %
  • Sulfate (SO4):
    max. 0.002 %
  • Trace Impurities (in ppm)Iron (Fe):
    max. 2
  • Trace Impurities (in ppm)Magnesium (Mg):
    max. 5
  • Trace Impurities (in ppm)Heavy Metals (as Pb):
    max. 5
  • Trace Impurities (in ppm)Phosphate (PO4):
    max. 2

 

 

Frequently Bought Together