


Cobalt(II) isopropoxide
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60406-95-3
:
Formula:C₆H₁₄CoO₂
MDL Number:MFCD00799075
ADR:4.1,III
:ampouled under argon
:178.11 g/mol
UN:3181
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together