(Methoxycarbonylmethyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1779-58-4
:
Formula:C₂₁H₂₀BrO₂P
MDL Number:MFCD00011801
:415.28 g/mol
Melting Pt:∼ 163 °C (dec.)
EINECS:217-222-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together