
2-Bromo-4'-(1-pyrrolidinyl)acetophenone ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:216144-18-2
:
Formula:C₁₂H₁₄BrNO
MDL Number:MFCD01075690
:268.16 g/mol
Melting Pt:120…122 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together