Order Entry
Czech Republic
ContactUsLinkComponent
4-Methoxy-2-methylphenylboronic acid, Sigma-Aldrich®
4-Methoxy-2-methylphenylboronic acid, Sigma-Aldrich®
Catalog # SIAL639370-1G
Supplier:  Merck
CAS Number:  
4-Methoxy-2-methylphenylboronic acid, Sigma-Aldrich®
Catalog # SIAL639370-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :165.99 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:208399-66-0
Formula:C₈H₁₁BO₃
MDL Number:MFCD02684315