Methylamine 2 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:74-89-5
:
Formula:CH₅N
MDL Number:MFCD00008104
:31.06 g/mol
Storage Temperature:Ambient
EINECS:200-820-0
UN:2924
ADR:3,II
Frequently Bought Together