Order Entry
Czech Republic
ContactUsLinkComponent
Potassium acetate 5 mol/l (5 N) in aqueous solution
Potassium acetate 5 mol/l (5 N) in aqueous solution
Catalog # ROCKMB-042
CAS Number:  
undefined
Potassium acetate 5 mol/l (5 N) in aqueous solution
Catalog # ROCKMB-042
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :98.14 g/mol
Storage Temperature:Ambient
EINECS:204-822-2
CAS Number:127-08-2
Formula:CH₃COOK
MDL Number:MFCD00012458