3'-Brom-4'-methyloacetophenone, Sigma-Aldrich®
Catalog # SIAL579734-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:213.0700 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:40180-80-1
Formula:C₉H₉BrO
MDL Number:MFCD02683854