Order Entry
Czech Republic
ContactUsLinkComponent
Methyl pentafluorobenzoate, Sigma-Aldrich®
Methyl pentafluorobenzoate, Sigma-Aldrich®
Catalog # SIAL303070-5G
Supplier:  Merck
CAS Number:  
Methyl pentafluorobenzoate, Sigma-Aldrich®
Catalog # SIAL303070-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :226.1002 g/mol
CAS Number:36629-42-2
Formula:C₈H₃F₅O₂
MDL Number:MFCD00012172