

4-(Dimethylamino)benzoic anhydride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7474-31-9
:
Formula:C₁₈H₂₀N₂O₃
MDL Number:MFCD09757554
:312.37 g/mol
Melting Pt:149…154 °C
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together