Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3',5'-Dinitroacetophenone
3',5'-Dinitroacetophenone, Sigma-Aldrich®
3',5'-Dinitroacetophenone, Sigma-Aldrich®
SIAL631299-1G
:
Merck
:
3',5'-Dinitroacetophenone, Sigma-Aldrich®
SIAL631299-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
M-Clarity™ quality level = MQ100
:
210.14 g/mol
:
14401-75-3
:
C₈H₆N₂O₅
:
MFCD00631987