Order Entry
Czech Republic
ContactUsLinkComponent
3-Bromo-4-fluoro-5-iodobenzaldehyde ≥97%
Irritant
3-Bromo-4-fluoro-5-iodobenzaldehyde ≥97%
Catalog # H33441.03
CAS Number:  
Irritant
3-Bromo-4-fluoro-5-iodobenzaldehyde ≥97%
Catalog # H33441.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :328.90 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1356113-37-5
Formula:C₇H₃BrFIO
MDL Number:MFCD20486165