
4-(N,N-Diphenylamino)phenylboronic acid 98%
Catalog # H52902.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
Specifications
About this item
CAS Number:201802-67-7
:
Formula:C₁₈H₁₆BNO₂
MDL Number:MFCD06798117
:289.14 g/mol
Melting Pt:110…115 °C
:GHS07
Storage Temperature:Ambient