0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Sodium acetate-[1-13C]
Sodium acetate-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
Sodium acetate-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
SIAL279293-1G
:
Merck
:
Sodium acetate-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
SIAL279293-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
M-Clarity™ quality level = MQ200
:
Ambient
:
23424-28-4
:
H₃CCOONa
:
MFCD00037666