Order Entry
Czech Republic
ContactUsLinkComponent
Nickel standard dissolved in oil, 1000 µg/g Ni in mineral oil for wear metal analysis, Specpure®
Nickel standard dissolved in oil, 1000 µg/g Ni in mineral oil for wear metal analysis, Specpure®
Catalog # 14211.18
CAS Number:  
Nickel standard dissolved in oil, 1000 µg/g Ni in mineral oil for wear metal analysis, Specpure®
Catalog # 14211.18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00011137
  • Element:
    Nickel
  • Symbol:
    Ni

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00011137