
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69304-37-6
:
Formula:C₁₂H₂₈Cl₂OSi₂
MDL Number:MFCD00009655
:315.43 g/mol
Boiling Pt:69…70 °C (0.5 mmHg)
Density:0.986
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:1760
ADR:8,II
:GHS05
Related Information
Frequently Bought Together