Order Entry
Czech Republic
ContactUsLinkComponent
 
 
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 96%
 
undefined
 
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 96%

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4533 to 1.4553 (20°C, 589 nm)
  • GC:
    ≥95 %

 

 

Frequently Bought Together