Bis[N,N-bis(trimethylsilyl)amide]zinc(II), Sigma-Aldrich®
Catalog # SIAL417696-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
CAS Number:14760-26-0
Formula:C₁₂H₃₆N₂Si₄Zn