Order Entry
Czech Republic
ContactUsLinkComponent
Bis[N,N-bis(trimethylsilyl)amide]tin(II), Sigma-Aldrich®
Bis[N,N-bis(trimethylsilyl)amide]tin(II), Sigma-Aldrich®
Catalog # SIAL409618-25ML
Supplier:  Merck
CAS Number:  
Bis[N,N-bis(trimethylsilyl)amide]tin(II), Sigma-Aldrich®
Catalog # SIAL409618-25ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
CAS Number:59863-13-7
Formula:C₁₂H₃₆N₂Si₄Sn
MDL Number:MFCD00191794