Silver standard dissolved in oil, 5,000 µg/g Ag in mineral oil for wear metal analysis, Specpure®
Catalog # 43868.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:5000 µg/g
- Matrix:Mineral oil
- Pack type:Glass bottle for solids
- Pk:50 g
- Description:Standard for wear metal analysis
- MDL No.:MFCD00003397
- Element:Silver
- Symbol:Ag
Specifications
About this item
Physical Form: Liquid
MDL No.: MFCD00003397