Silver standard dissolved in oil, 1000 µg/g Ag in mineral oil for wear metal analysis, Specpure®
Catalog # 14199.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:1000 µg/g
- Matrix:oil
- Pack type:Glass bottle for solids
- Pk:50 g
- Description:Standard for wear metal analysis
- MDL No.:MFCD00003397
- Element:Silver
- Symbol:Ag
Specifications
About this item
Physical form: liquid
MDL No.: MFCD00003397