Order Entry
Czech Republic
ContactUsLinkComponent
2,4-Dimethoxy-5-pyrimidineboronic acid, Sigma-Aldrich®
2,4-Dimethoxy-5-pyrimidineboronic acid, Sigma-Aldrich®
Catalog # SIAL666491-1G
Supplier:  Merck
CAS Number:  
2,4-Dimethoxy-5-pyrimidineboronic acid, Sigma-Aldrich®
Catalog # SIAL666491-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.9577 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:89641-18-9
Formula:C₆H₉BN₂O₄
MDL Number:MFCD01318985