Order Entry
Czech Republic
ContactUsLinkComponent
 
Aluminium standard dissolved in oil, 1000 µg/g Al in Base Oil 20
Catalog # PROOCON150-100-135
CAS Number:  
Aluminium standard dissolved in oil, 1000 µg/g Al in Base Oil 20
Catalog # PROOCON150-100-135
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Base Oil 20
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item