1-Boc-3-(methylamino)piperidine
Catalog # ABCRNHC-3123.0001
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:214.31 g/mol
CAS Number:392331-89-4
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD04114980