Order Entry
Czech Republic
ContactUsLinkComponent
Bromotrispyrrolidinophosphonium hexafluorophosphate 97%
Irritant
Bromotrispyrrolidinophosphonium hexafluorophosphate 97%
Certificates
undefined
Irritant
Bromotrispyrrolidinophosphonium hexafluorophosphate 97%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    ≥100 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together