Order Entry
Czech Republic
ContactUsLinkComponent
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide for synthesis, Sigma-Aldrich®
Irritant
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide for synthesis, Sigma-Aldrich®
Catalog # 8.18094.0010
Supplier:  Merck
CAS Number:  
Irritant
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide for synthesis, Sigma-Aldrich®
Catalog # 8.18094.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    194 - 196 °C
  • Molar Mass:
    429.29 g/mol

Specification Test Results

About this item

 :
 :429.30 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:257-977-3
 :GHS07
Storage Temperature:Ambient
CAS Number:52509-14-5
Formula:C₂₂H₂₂BrO₂P
MDL Number:MFCD00011966