
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52509-14-5
:
Formula:C₂₂H₂₂BrO₂P
MDL Number:MFCD00011966
:429.30 g/mol
Melting Pt:186…189 °C
EINECS:257-977-3
:GHS07
Related Information
Frequently Bought Together