Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Carbamazepine 100 µg/ml in acetonitrile, CIL
Organic reference standard, Carbamazepine 100 µg/ml in acetonitrile, CIL
Catalog # PROOCILULM65811.2
CAS Number:  
Organic reference standard, Carbamazepine 100 µg/ml in acetonitrile, CIL
Catalog # PROOCILULM65811.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    298-46-4
  • Description:
    Carbamazepine 100 µg/ml in acetonitrile
  • Formula:
    C₁₅H₁₂N₂O
  • Description:
    Carbamazepine

Specifications

About this item