Buffer, TRIS solution pH 8,0 (2 mol/L), Sigma-Aldrich®
: Merck
- M-Clarity quality level:MQ200
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® Blender Bags
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Petri dishes
: GAMA GROUP
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
VWR® Standard Line Transfer Pipettes
: VWR Collection