Order Entry
Czech Republic
ContactUsLinkComponent
 
Buffers, TRIS, 1,0 M solutions
 
Buffers, TRIS, 1,0 M solutions
 

 

  • CAS No.:
    77-86-1
  • MDL No.:
    MFCD00004679
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Appearance:
    Colourless

 

Frequently Bought Together