Order Entry
Czech Republic
ContactUsLinkComponent
 
Buffers, sodium bicarbonate, 1 M solutions
 
Buffers, sodium bicarbonate, 1 M solutions
 

 

  • CAS No.:
    144-55-8
  • MDL No.:
    MFCD00003528
  • Physical form:
    Liquid

 

Frequently Bought Together