Order Entry
Czech Republic
ContactUsLinkComponent
Buffer, BIS-TRIS propane, 0,2 M solution, pH 6,5
Buffer, BIS-TRIS propane, 0,2 M solution, pH 6,5
Certificates
Buffer, BIS-TRIS propane, 0,2 M solution, pH 6,5
Certificates

Specifications

  • CAS No.:
    64431-96-5
  • MDL No.:
    MFCD00004689
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together