



(Trimethylsilyl)diazomethane 2 M in hexanes
Catalog # ABCRAB212318.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
Density:0.718
Flash Pt:–23 °C (–10 °F)
ADR:6.1,I
:114.22 g/mol
Storage Temperature:Ambient
UN:3383
:GHS02
:GHS06
:GHS08
:GHS07
CAS Number:18107-18-1
Formula:C₄H₁₀N₂Si
MDL Number:MFCD00053946