4-(1-Pyrrolidinyl)pyridine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2456-81-7
:
Formula:C₉H₁₂N₂
MDL Number:MFCD00006415
:148.21 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:219-534-2
UN:2923
Frequently Bought Together