ContactUsLinkComponent
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Irritant
Health
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Certificates
undefined
Irritant
Health
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Specification Test Results
  • Appearance:
    Orange to red-brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Carbon (C):
    50 to 53 %
  • Palladium (Pd):
    12.6 to 13.4 %