[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
Catalog # ABCRAB152323.0010
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:731.70 g/mol
EINECS:460-040-3
CAS Number:72287-26-4
Formula:C₃₄H₂₈Cl₂P₂FePd
MDL Number:MFCD00015757