[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:67292-34-6
:
Formula:C₃₄H₂₈Cl₂FeNiP₂
MDL Number:MFCD00270284
:683.98 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together