Organic reference standard, Musk ketone 100 µg/ml in isooctane
Catalog # PROOSL20884
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- CAS No.:81-14-1
- Description:Musk ketone 100 µg/ml in isooctane
- Formula:C₁₄H₁₈N₂O₅
- Description:Musk ketone