HPLC columns, Hypersil GOLDâ˘
These Hypersil GOLD⢠HPLC columns are ideal for LS/MS applications to achieve exceptional peak shape and resolution.
- Endcapped and ultrapure
- Silica-based columns with exceptional peak shape and resolution
- Rugged sleeved design for robustness
- High performance and sensitivity
These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give you confidence in the accuracy and quality of your analytical data. 1,9 Âľm particle size columns can improve the speed and efficiency of your applications.
: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ă require the 1 mm UNIGUARD holder, 2,1/3 mm Int.Ă require the 2/3 mm UNIGUARD holder and 4/4,6 mm Int.Ă require the 4/4,6 mm UNIGUARD holder.
- Catalog No:
- 555-8052
- 555-0145
- 555-7695
- 555-8050
- 555-8007
- 555-8002
- 555-0152
- 555-0146
- 555-0153
- 555-8048
- 555-0151
- 555-8133
- 555-0149
- 555-0143
- 555-0150
- 555-0144
- 555-0141
- 555-0147
- 555-0142
- 555-0148
- 555-0140
- 555-0154
- 555-0155
- Carbon load:
- 10%
- 10%
- 10%
- 10%
- pH range:
- 1 - 11
- 1 to 11
- 1 - 11
- 1 - 11
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
Frequently Bought Together