Order Entry
Czech Republic
ContactUsLinkComponent
 
Affinity chromatography column, GSTPrep™ FF
  28-9365-50
 :  Cytiva
undefined
Affinity chromatography column, GSTPrep™ FF
  28-9365-50
 :  Cytiva

 

  • Flow rate:
    <10 ml/min
  • Particle Size:
    90 µm
  • Max. pressure:
    1,5 bar
  • Capacity:
    20 ml
  • Description:
    GSTPrep™ FF 16/10
  • Seq:
    01
  • pH range:
    3 - 12
  • Storage:
    4 to 30 °C, 20% Ethanol
  • Column hardware:
    PP
  • Bed dimensions:
    16×100 mm
  • Flow velocity:
    <300 cm/h
  • Pk:
    1

 

 

GSTPrep™ FF 16/10 is prepacked, ready-to-use column containing glutathione Sepharose™ 4 fast flow designed for one-step, scale-up purification of glutathione S-transferase (GST)-tagged proteins.

  • Binds approximately 200 mg recombinant GST/column
  • Mild elution conditions preserving protein antigenicity and function
  • Simple operation with a pump or chromatography system, such as ÄKTA design

Glutathione Sepharose™ 4 fast flow is for scale-up purification of recombinant GST-tagged proteins, other glutathione S-transferases, and glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function.