ContactUsLinkComponent
Anion exchange chromatography columns, HiTrap™ ANX Sepharose™ FF
Anion exchange chromatography columns, HiTrap™ ANX Sepharose™ FF
Supplier:  Cytiva
Certificates
undefined
Anion exchange chromatography columns, HiTrap™ ANX Sepharose™ FF
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 17-5162-01
  • 17-5163-01
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 13
  • 3 - 13