Order Entry
Czech Republic
ContactUsLinkComponent
 
Columns, PD MiniTrap™ G-10
Catalog # 28-9180-10
Supplier:  Cytiva
undefined
Columns, PD MiniTrap™ G-10
Catalog # 28-9180-10
Supplier:  Cytiva

Specifications

  • Capacity:
    2,1 ml
  • Description:
    PD MiniTrap™ G-10
  • Column hardware:
    PP
  • Bed dimensions:
    9,7×28 mm
  • Pk:
    50

Specifications

About this item

PD MiniTrap G-10 columns are prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides.

  • Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700 before downstream analysis such as liquid chromatography, electrophoresis, and MS
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels
  • High desalting capacity
  • Preparation of samples in the volume range of 100 to 300 μl

PD MiniTrap G-10 gravity columns give fast and simple desalting and buffer exchange without any need for a purification system. PD MiniTrap G-10 columns are a member of the Trap platform, which addresses the need for flexible, small-scale preparation of proteins and peptides before analysis.

For larger scale please use PD MidiTrap G-10 columns.

Delivery: Instructions are supplied with the columns.