ContactUsLinkComponent
Proteome kits, mammalian proteome, FOCUS™
Proteome kits, mammalian proteome, FOCUS™
Catalog # 786-246
Supplier:  G-Biosciences
CAS Number:  
undefined
Proteome kits, mammalian proteome, FOCUS™
Catalog # 786-246
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    FOCUS™ Mammalian Proteome, 50 preps
Specifications
About this item
Extracts and solubilises nearly all of the proteins from mammalian samples, including membrane as well as soluble proteins, by a strong chaotropic extraction buffer to solubilise even the most difficult proteins. Suitable for biological samples from animal tissues and adherent and suspension cells.
  • Single step extraction protocol
  • Supplied with a strong proprietary chaotropic extraction buffer
  • For 50×100 mg animal tissue or 50 μl wet cell pellet preparations
  • Suitable for sample preparation for 2D gel electrophoresis and other applications