2',3'-Dideoxyinosine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69655-05-6
:
Formula:C₁₀H₁₂N₄O₃
MDL Number:MFCD00077728
Merck Index:14,3098
:236.23 g/mol
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Antiviral compounds
Product Literature
Frequently Bought Together