L(+)-Ornithine hydrochloride cell culture reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3184-13-2
:
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00064562
:168.62 g/mol
Merck Index:14,6874
Melting Pt:∼ 238 °C (dec.)
Storage Temperature:Ambient
EINECS:221-678-6
Related Information
Frequently Bought Together