Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Sodium hydroxide ≥98% (by acidimetry), pellets, AR® ACS, Macron Fine Chemicals™
 
undefined
 
Sodium hydroxide ≥98% (by acidimetry), pellets, AR® ACS, Macron Fine Chemicals™
 

 

  • For Laboratory,Research,or Manufacturing Use:
    = 98 %
  • Assay (NaOH) (by acidimetry):
    ≥98 %
  • Identification:
    Passes Test
  • Calcium (Ca):
    ≤0.005 %
  • Chloride (Cl):
    ≤0.005 %
  • Copper (Cu):
    ≤0.001 %
  • Heavy Metals (as Ag):
    ≤0.001 %
  • Insoluble Matter:
    ≤0.003 %
  • Iron (Fe):
    ≤0.0003 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • Mercury (Hg):
    ≤0.1 ppm
  • Nickel (Ni):
    ≤0.0005 %
  • Nitrogen Compounds (as N):
    ≤0.0003 %
  • Phosphate (PO₄):
    ≤0.0002 %
  • Potassium (K):
    ≤0.02 %
  • Sodium Carbonate (Na₂CO₃):
    ≤0.4 %
  • ACS - Sulfate (SO₄):
    ≤0.003 %

 

 

Frequently Bought Together