Order Entry
Czech Republic
ContactUsLinkComponent
1,3-Dihydroxyacetone dimer
1,3-Dihydroxyacetone dimer
Catalog # J60592.06
CAS Number:  
1,3-Dihydroxyacetone dimer
Catalog # J60592.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:96-26-4
 :
Formula:C₃H₆O₃
 :180.16 g/mol
Merck Index:14,3182
Melting Pt:75…80 °C
Density:1.52 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:202-494-5
CAS Number:62147-49-3
Formula:C₆H₁₂O₆
MDL Number:MFCD00051019