
1,4-Dithioerythritol 99% for electrophoresis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6892-68-8
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00063750
:154.25 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:82…84 °C
Storage Temperature:Refrigerator
EINECS:229-998-8
UN:3335
ADR:9,
:GHS07
Related Information
Frequently Bought Together